Bradykinin
Supplier: MilliporeSigma
05-23-0500-1MG 05-23-0500-5MG
80011-666EA 27.94 USD
80011-666 80603-870
Bradykinin
Proteins and Peptides
Bradykinin, Induces the release of nitric oxide.
Bradykinin, a nonapeptide, is produced in plasma under inflammatory conditions by cleavage of kininogen by kallikrein. Bradykinin is involved in a number of physiological functions, including the release of nitric oxide, stimulation of pain receptors, induction of antimitogenic effects, inhibition of cAMP accumulation, induction of smooth muscle contraction, and vasodilation, as well as in inflammation processes. Also involved in edema resulting from trauma or injury. Improves post-ischemic recovery of the heart via a nitric oxide-dependent mechanism.