[5-(Aminomethyl)furan-2-yl]methanol, 95%
Supplier: Ambeed
|
Danger
|
A1011979-100MG
A1011979-1G
A1011979-250MG
77782-848EA
150
USD
77782-848
77782-849
77782-850
[5-(Aminomethyl)furan-2-yl]methanol, 95%
[5-(Aminomethyl)furan-2-yl]methanol
Precaución:
for Research Use Only
Formula:
C₆H₉NO₂ |
MDL Number:
MFCD15145882 CAS Number: 88910-22-9 UN: 2735 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...