tert-Butyl (cis-3-(aminomethyl)cyclobutyl)carbamate, 98%

Supplier: Ambeed

A161097-100MG A161097-250MG
77685-324EA 125 USD
77685-324 77685-325
tert-Butyl (cis-3-(aminomethyl)cyclobutyl)carbamate, 98%
tert-Butyl N-[cis-3-(aminomethyl)cyclobutyl]carbamate

Precaución:

for Research Use Only


Formula: C₁₀H₂₀N₂O₂
MDL Number: MFCD20921511
CAS Number: 871014-28-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR