1,4-Dibromo-2-(bromomethyl)benzene, 95%

Supplier: Ambeed
Danger

A987899-25G A987899-5G
77784-597EA 124 USD
77784-597 77784-598
1,4-Dibromo-2-(bromomethyl)benzene, 95%
1,4-Dibromo-2-(bromomethyl)benzene

Precaución:

for Research Use Only


Formula: C₇H₅Br₃
MDL Number: MFCD11052870
CAS Number: 136105-40-3
UN: 3261

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR