2-(2-(2-Hydroxyethoxy)ethoxy)acetic acid 95%

Supplier: Ambeed
Danger

A754412-250MG A754412-100MG
77783-914EA 62 USD
77783-914 77783-913
2-(2-(2-Hydroxyethoxy)ethoxy)acetic acid 95%
Hydroxy-PEG2-CH2CO2H

Precaución:

for Research Use Only


Formula: C₆H₁₂O₅
MDL Number: MFCD20640451
CAS Number: 51951-04-3
UN: 3265

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR