2-Oxa-8-azaspiro[4.5]decan-1-one, 97%

Supplier: Ambeed
Warning

A1151147-100MG A1151147-1G A1151147-250MG A1151147-5G
77865-980EA 73 USD
77865-980 77865-981 77865-982 77865-983
2-Oxa-8-azaspiro[4.5]decan-1-one, 97%
2-Oxa-8-azaspiro[4.5]decan-1-one

Precaución:

for Research Use Only


Formula: C₈H₁₃NO₂
MDL Number: MFCD11847703
CAS Number: 4427-25-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR