2',5'-Difluoro-[1,1':4',1''-terphenyl]-3,3'',5,5''-tetracarboxylic acid, 97%

Supplier: Ambeed
Warning

A1351834-100MG A1351834-250MG
77867-437EA 87 USD
77867-437 77867-438
2',5'-Difluoro-[1,1':4',1''-terphenyl]-3,3'',5,5''-tetracarboxylic acid, 97%
2',5'-Difluoro-[1,1':4',1''-terphenyl]-3,3'',5,5''-tetracarboxylic acid

Precaución:

for Research Use Only


Formula: C₂₂H₁₂F₂O₈
CAS Number: 1119195-97-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR