Ethyl 3,4-difluoro-5-hydroxybenzoate 95%

Supplier: Ambeed

A470886-5G A470886-250MG A470886-25G A470886-1G A470886-10G
77748-975EA 195 USD
77748-975 77748-973 77748-974 77748-972 77748-971
Ethyl 3,4-difluoro-5-hydroxybenzoate 95%
Ethyl 3,4-difluoro-5-hydroxybenzoate

Precaución:

for Research Use Only


Formula: C₉H₈F₂O₃
MDL Number: MFCD11042806
CAS Number: 847943-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR