1-Bromo-4-(trifluoromethyl)cyclohexane, 98%

Supplier: Ambeed
Danger

A864567-100MG A864567-1G A864567-250MG
77784-262EA 148 USD
77784-262 77784-263 77784-264
1-Bromo-4-(trifluoromethyl)cyclohexane, 98%
1-Bromo-4-(trifluoromethyl)cyclohexane

Precaución:

for Research Use Only


Formula: C₇H₁₀BrF₃
MDL Number: MFCD20410249
CAS Number: 30129-20-5
UN: 1993

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR