tert-Butyl 3,8-diazabicyclo[3.2.1]octane-8-carboxylate hydrochloride, 97%

Supplier: Ambeed

A524406-1G A524406-250MG A524406-100MG
77749-094EA 154 USD
77749-094 77749-095 77749-093
tert-Butyl 3,8-diazabicyclo[3.2.1]octane-8-carboxylate hydrochloride, 97%
tert-Butyl 3,8-diazabicyclo[3.2.1]octane-8-carboxylate hydrochloride

Precaución:

for Research Use Only


Formula: C₁₁H₂₁ClN₂O₂
MDL Number: MFCD23381425
CAS Number: 1403676-97-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR