1-((1R,2R)-2-Aminocyclohexyl)-3-(3,5-bis(trifluoromethyl)phenyl)thiourea 98%

Supplier: Ambeed

A180917-100MG A180917-250MG
77746-176EA 104 USD
77746-176 77746-177
1-((1R,2R)-2-Aminocyclohexyl)-3-(3,5-bis(trifluoromethyl)phenyl)thiourea 98%
1-((1R,2R)-2-Aminocyclohexyl)-3-(3,5-bis(trifluoromethyl)phenyl)thiourea

Precaución:

for Research Use Only


Formula: C₁₅H₁₇F₆N₃S
MDL Number: MFCD30541720
CAS Number: 860994-58-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR