13,13-Dimethyl-11-oxo-3,6,9,12-tetraoxatetradecan-1-oic acid 97%

Supplier: Ambeed
Warning

A1004840-250MG A1004840-5G
77782-752EA 52 USD
77782-752 77782-753
13,13-Dimethyl-11-oxo-3,6,9,12-tetraoxatetradecan-1-oic acid 97%
13,13-Dimethyl-11-oxo-3,6,9,12-tetraoxatetradecan-1-oic acid

Precaución:

for Research Use Only


Formula: C₁₂H₂₂O₇
MDL Number: MFCD31715330
CAS Number: 883564-93-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR