tert-Butyl 4-(4-aminobutyl)piperidine-1-carboxylate 98%

Supplier: Ambeed
Warning

A1213507-100MG A1213507-1G A1213507-250MG
77866-690EA 125 USD
77866-690 77866-691 77866-692
tert-Butyl 4-(4-aminobutyl)piperidine-1-carboxylate 98%
tert-Butyl 4-(4-aminobutyl)piperidine-1-carboxylate

Precaución:

for Research Use Only


Formula: C₁₄H₂₈N₂O₂
MDL Number: MFCD09913671
CAS Number: 201034-98-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR