(1S)-1-Cyclobutylethan-1-amine hydrochloride 97%

Supplier: Ambeed

A248250-100MG A248250-1G A248250-250MG A248250-5G
77748-457EA 199 USD
77748-457 77748-458 77748-459 77748-460
(1S)-1-Cyclobutylethan-1-amine hydrochloride 97%
(1S)-1-Cyclobutylethan-1-amine hydrochloride

Precaución:

for Research Use Only


Formula: C₆H₁₄ClN
MDL Number: MFCD27980977
CAS Number: 1355969-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR