tert-Butyl 2-(hydroxymethyl)-7-azaspiro[3.5]nonane-7-carboxylate, 98%

Supplier: Ambeed
Warning

A241836-100MG A241836-250MG
77685-674EA 31 USD
77685-674 77685-675
tert-Butyl 2-(hydroxymethyl)-7-azaspiro[3.5]nonane-7-carboxylate, 98%
7-Boc-7-azaspiro[3.5]nonane-2-methanol

Precaución:

for Research Use Only


Formula: C₁₄H₂₅NO₃
MDL Number: MFCD20926183
CAS Number: 1356476-27-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR