2-Ethynylpyrrolidine hydrochloride 98%

Supplier: Ambeed

A1147224-100MG A1147224-1G
77865-905EA 85 USD
77865-905 77865-906
2-Ethynylpyrrolidine hydrochloride 98%
2-Ethynylpyrrolidine hydrochloride

Precaución:

for Research Use Only


Formula: C₆H₁₀ClN
MDL Number: MFCD15143301
CAS Number: 1314937-87-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR