Anti-HIST1H1B Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A24655-100UL A24655-50UL
77739-708EA 350 USD
77739-708 77856-871
Anti-HIST1H1B Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB: 1:100 to 1:1000, IHC: 1:20 to 1:200

Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR