Anti-CDH-1 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A50178-100UG A50178-50UG
77814-796EA 330 USD
77814-796 77814-797
Anti-CDH-1 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: CDH-1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Phanerochaete chrysosporium
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR