Anti-L-lactate dehydrogenase Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A56074-100UG A56074-50UG
77825-590EA 330 USD
77825-590 77825-591
Anti-L-lactate dehydrogenase Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: L-lactate dehydrogenase
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Plasmodium falciparum
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR