Anti-METTL14 Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A70278-100UG A70278-50UG
77842-197EA 330 USD
77842-197 77842-198
Anti-METTL14 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: METTL14
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR