Anti-Larval cuticle protein A1A Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A50093-100UG A50093-50UG
77814-642EA 330 USD
77814-642 77814-643
Anti-Larval cuticle protein A1A Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: Larval cuticle protein A1A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Tenebrio molitor
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR