Anti-Arginine esterase Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A51664-100UG A51664-50UG
77817-601EA 330 USD
77817-601 77817-602
Anti-Arginine esterase Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: Arginine esterase
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR