Anti-SLC25A52 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A56399-100UG A56399-50UG
77826-204EA 330 USD
77826-204 77826-205
Anti-SLC25A52 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: SLC25A52
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR