Anti-H2AFY2 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A70474-100UG A70474-50UG
77842-566EA 330 USD
77842-566 77842-567
Anti-H2AFY2 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: H2AFY2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR