Anti-Ribosome-inactivating protein PD-L1/PD-L2 Rabbit Recombinant Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: AFG Bioscience
A50114-50UG A50114-100UG
77814-676EA 225 USD
77814-676 77814-675
Anti-Ribosome-inactivating protein PD-L1/PD-L2 Rabbit Recombinant Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Type:
Antigen: Ribosome-inactivating protein PD-L1/PD-L2
Clonality: Recombinant
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Phytolacca dioica
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR