Anti-dye Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A59664-100UG A59664-50UG
77832-321EA 330 USD
77832-321 77832-322
Anti-dye Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: dye
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR