Anti-VDAC2 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A38276-100UG A38276-50UG
77804-317EA 330 USD
77804-317 77804-318
Anti-VDAC2 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC: 1:20 to 1:200

Type:
Antigen: VDAC2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR