Anti-RSPO3 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A70685-100UG A70685-50UG
77842-969EA 330 USD
77842-969 77842-970
Anti-RSPO3 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: RSPO3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR