Anti-TBKBP1 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A36219-100UL A36219-50UL
77801-954EA 330 USD
77801-954 77801-955
Anti-TBKBP1 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: TBKBP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR