Anti-HLA-F Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A50864-100UG A50864-50UG
77816-096EA 330 USD
77816-096 77816-097
Anti-HLA-F Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IF: 1:50 to 1:200

Type:
Antigen: HLA-F
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR