p-Tolylacetonitrile ≥98.0%
Supplier: TCI America
CAS Number: 2947-61-7
MDL Number: MFCD00001922
Molecular Formula: C9H9N
Molecular Weight: 131.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 245
Flash Point (°C): 106
Freezing point (°C): 15
Specific Gravity (20/20): 0.99
Other Product Names / Synonyms: 4-Methylbenzyl Cyanide, p-Tolylacetonitrile
Formula:
C₉H₉N Boiling Pt: 245 °C Melting Pt: 18 °C Density: 0.99 (20 °C) Flash Pt: 106 °C |
MDL Number:
MFCD00001922 CAS Number: 2947-61-7 EINECS: 220-963-2 UN: 3276 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...