m-Xylene ≥99.0%

Supplier: TCI America
Danger

X0013-500ML X0013-25ML
TCX0013-500MLEA 56.5 USD
TCX0013-500ML TCX0013-25ML
m-Xylene ≥99.0%
m-Xylene

CAS Number: 108-38-3
MDL Number: MFCD00008536
Molecular Formula: C8H10
Molecular Weight: 106.17
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 139
Flash Point (°C): 28
Specific Gravity (20/20): 0.87


Other Product Names / Synonyms: 1,3-Dimethylbenzene

Formula: C₈H₁₀
Boiling Pt: 139 °C
Melting Pt: –48 °C
Density: 0.87 (20 °C)
Flash Pt: 28 °C
Storage Temperature: Ambient
MDL Number: MFCD00008536
CAS Number: 108-38-3
EINECS: 203-576-3
UN: 1307
ADR: 3,III
Merck Index: 12,10214
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d07b9fa8c6b8f296369d7c5dc4556f6adfea8412&vid=ce22e797390cc83a8d50ffbdf0f55436d536ac3d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR