2-Vinylpyridine ≥97.0% stabilized

Supplier: TCI America


Stabilization: Stabilised with Stabilised

V0024-500ML V0024-25ML
TCV0024-500MLEA 164 USD
TCV0024-500ML TCV0024-25ML
2-Vinylpyridine ≥97.0% stabilized
2-Vinylpyridine

(stabilized with TBC)
CAS Number: 100-69-6
MDL Number: MFCD00006355
Molecular Formula: C7H7N
Molecular Weight: 105.14
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 158
Melting point (°C): -50
Flash Point (°C): 46
Specific Gravity (20/20): 0.98
Storage Temperature: <0°C

Formula: C₇H₇N
MW: 105.14 g/mol
Boiling Pt: 158 °C
Melting Pt: -50 °C
Density: 0.98 (20 °C)
Flash Pt: 46 °C
Storage Temperature: Freezer
MDL Number: MFCD00006355
CAS Number: 100-69-6
EINECS: 202-879-8
UN: 3073
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1d6c5a94391def261b11e9fbe1b1c8bdfd80f35&vid=e1d6c5a94391def261b11e9fbe1b1c8bdfd80f35&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR