Valeryl chloride ≥98.0%

Supplier: TCI America

Synonyms: Pentanoyl chloride, Valeroyl chloride

V0010-25ML V0010-100ML
TCV0010-25MLEA 31.53 USD
TCV0010-25ML TCV0010-100ML
Valeryl chloride ≥98.0%
Valeryl chloride

CAS Number: 638-29-9
MDL Number: MFCD00000757
Molecular Formula: C5H9ClO
Molecular Weight: 120.58
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 126
Melting point (°C): -110
Flash Point (°C): 32
Specific Gravity (20/20): 1.00


Other Product Names / Synonyms: Valeroyl Chloride

Formula: C₅H₉ClO
MW: 120.58 g/mol
Boiling Pt: 125
Melting Pt: -110
Density: 1.00 (20 °C)
Flash Pt: 32 °C
MDL Number: MFCD00000757
CAS Number: 638-29-9
EINECS: 211-330-1
UN: 2502
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0772b4d6d8d305a7feb2ee1145fffbb3221001de&vid=0772b4d6d8d305a7feb2ee1145fffbb3221001de&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR