(±)-2-Undecanol ≥98.0%

Supplier: TCI America

Synonyms: Undecan-2-ol, (±)-Methyl nonyl carbinol, (±)-2-Hydroxy undecane, (±)-Nonyl methyl carbinol, 2-Undecanol

U0027-5ML U0027-25ML
TCU0027-005MLEA 31 USD
TCU0027-005ML TCU0027-025ML
(±)-2-Undecanol ≥98.0%
(±)-2-Undecanol

CAS Number: 1653-30-1
MDL Number: MFCD00021958
Molecular Formula: C11H24O
Molecular Weight: 172.31
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 228
Melting point (°C): 3
Flash Point (°C): 113
Specific Gravity (20/20): 0.83

Formula: C₁₁H₂₄O
MW: 172.31 g/mol
Boiling Pt: 228 °C
Melting Pt: 3 °C
Density: 0.83 (20 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00021958
CAS Number: 1653-30-1
EINECS: 216-722-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=37620d438f4a09140b492f5fd7ee167d654e0a25&vid=dab976921e9631af4e6a5f316d00e9eb8e458459&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR