Undecanal ≥97.0%
Supplier: TCI America
Synonyms:
Hendecanal, Undecylaldehyde
U0009-250ML
U0009-25ML
TCU0009-250MLEA
166
USD
TCU0009-250ML
TCU0009-025ML
Undecanal ≥97.0%
Undecanal
CAS Number: 112-44-7
MDL Number: MFCD00007016
Molecular Formula: C11H22O
Molecular Weight: 170.30
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 80
Melting point (°C): -2
Flash Point (°C): 113
Specific Gravity (20/20): 0.83
Other Product Names / Synonyms: Undecanaldehyde
Formula:
C₁₁H₂₂O MW: 170.3 g/mol Boiling Pt: 80 °C Melting Pt: –2 °C Density: 0.83 (20 °C) Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007016 CAS Number: 112-44-7 EINECS: 203-972-6 |
|
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...