Undecanoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

U0004-25G U0004-250G
TCU0004-025GEA 28.48 USD
TCU0004-025G TCU0004-250G
Undecanoic acid ≥98.0% (by GC, titration analysis)
Undecanoic acid

CAS Number: 112-37-8
MDL Number: MFCD00002730
Molecular Formula: C11H22O2
Molecular Weight: 186.30
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 250
Flash Point (°C): 110
Freezing point (°C): 28

Formula: C₁₁H₂₂O₂
Boiling Pt: 250 °C
Melting Pt: 30 °C
Density: 0.89 g/cm³ (20 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00002730
CAS Number: 112-37-8
EINECS: 203-964-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b620a963809d344acaf8e9d17fe1c22413158e71&vid=c9851e70750760e788f0c8574ff1076a03ea91b8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR