Potassium thioacetate ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Potassium ethanethioate

T2030-250G T2030-25G
TCT2030-250GEA 260.49 USD
TCT2030-250G TCT2030-25G
Potassium thioacetate ≥97.0% (by titrimetric analysis)
Potassium thioacetate

CAS Number: 10387-40-3
MDL Number: MFCD00083065
Molecular Formula: C2H4OS
Molecular Weight: 114.20
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Color: White
Melting point (°C): 176


Other Product Names / Synonyms: Thioacetic Acid S-Potassium Salt

Formula: C₂H₃KOS
MW: 114.21 g/mol
Melting Pt: 176 °C
MDL Number: MFCD00137704
CAS Number: 10387-40-3
EINECS: 233-848-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e71f02d25418cd0004fad02362c0938667d188f0&vid=e71f02d25418cd0004fad02362c0938667d188f0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR