Triclosan ≥96.0%

Supplier: TCI America

Synonyms: 5-Chloro-2-(2,4-dichlorophenoxy)phenol, 5-Chloro-2-(2, 4-dichlorophenoxy) phenol

T1872-500G T1872-25G T1872-100G
TCT1872-500GEA 312 USD
TCT1872-500G TCT1872-25G TCT1872-100G
Triclosan ≥96.0%
Triclosan

CAS Number: 3380-34-5
MDL Number: MFCD00800992
Molecular Formula: C12H7Cl3O2
Molecular Weight: 289.54
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 290
Melting point (°C): 57


Other Product Names / Synonyms: 5-Chloro-2-(2,4-dichlorophenoxy)phenol, Triclosan

Formula: C₁₂H₇Cl₃O₂
MW: 289.54 g/mol
Boiling Pt: 290 °C
Melting Pt: 57 °C
MDL Number: MFCD00800992
CAS Number: 3380-34-5
EINECS: 222-182-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c0f79b9dd37651096ee61c55f704b75a51e6d0a&vid=8c0f79b9dd37651096ee61c55f704b75a51e6d0a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR