Propyltrimethoxysilane ≥98.0%

Supplier: TCI America

Synonyms: Trimethoxy(propyl)silane

T1801-500ML T1801-25ML
TCT1801-500MLEA 101 USD
TCT1801-500ML TCT1801-25ML
Propyltrimethoxysilane ≥98.0%
Propyltrimethoxysilane

CAS Number: 1067-25-0
MDL Number: MFCD00043026
Molecular Formula: C6H16O3Si
Molecular Weight: 164.28
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 34
Specific Gravity (20/20): 0.94


Product Categories: Materials Chemistry, Silane Coupling Agents
Other Product Names / Synonyms: Propyltrimethoxysilane

Formula: C₆H₁₆O₃Si
MW: 164.28 g/mol
Boiling Pt: 142…143 °C (1013 hPa)
Density: 0.94 (20 °C)
Flash Pt: 34 °C
MDL Number: MFCD00043026
CAS Number: 1067-25-0
EINECS: 213-926-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da2df305dc92310635f8506e57151f3b4bec646c&vid=da2df305dc92310635f8506e57151f3b4bec646c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR