(α,α,α-Trifluoro-4-tolyl)acetic acid ≥96.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
(4-(Trifluoromethyl)phenyl)acetic acid, 4-(Trifluoromethyl)phenylacetic acid
T1783-25G
T1783-1G
T1783-5G
TCT1783-25GEA
165
USD
TCT1783-25G
TCT1783-001G
TCT1783-005G
(α,α,α-Trifluoro-4-tolyl)acetic acid ≥96.0% (by titrimetric analysis)
(α,α,α-Trifluoro-4-tolyl)acetic acid
CAS Number: 32857-62-8
MDL Number: MFCD00004352
Molecular Formula: C9H7F3O2
Molecular Weight: 204.15
Purity/Analysis Method: >96.0% (T)
Form: Crystal
Color: White
Melting point (°C): 83
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: (alpha,alpha,alpha-Trifluoro-p-tolyl)acetic Acid
Formula:
(C₆H₄(CF₃))CH₂COOH MW: 204.15 g/mol Melting Pt: 83 °C |
MDL Number:
MFCD00004352 CAS Number: 32857-62-8 EINECS: 251-263-5 |
Specification Test Results
Purity/Analysis Method | >96.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...