2,2,4,4-Tetramethylpentane ≥99.0%
Supplier: TCI America
|
Danger
|
T1704-25ML
T1704-5ML
TCT1704-025MLEA
208
USD
TCT1704-025ML
TCT1704-005ML
2,2,4,4-Tetramethylpentane ≥99.0%
2,2,4,4-Tetramethylpentane
CAS Number: 1070-87-7
MDL Number: MFCD00060873
Molecular Formula: C9H20
Molecular Weight: 128.26
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 122
Flash Point (°C): 16
Specific Gravity (20/20): 0.72
Formula:
C₉H₂₀ MW: 128.26 g/mol Boiling Pt: 122 °C Melting Pt: –66.5 °C Density: 0.72 (20 °C) Flash Pt: 16 °C Storage Temperature: Ambient |
MDL Number:
MFCD00060873 CAS Number: 1070-87-7 UN: 1920 ADR: 3,III |
|
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...