5-(Trifluoromethyl)uracil ≥95.0%
Supplier: TCI America
Synonyms:
Trifluorothymine, 5-(Trifluoromethyl)pyrimidine-2,4(1H,3H)-dione
T1622-5G
T1622-1G
TCT1622-5GEA
214
USD
TCT1622-5G
TCT1622-001G
5-(Trifluoromethyl)uracil ≥95.0%
5-(Trifluoromethyl)uracil
CAS Number: 54-20-6
MDL Number: MFCD00006024
Molecular Formula: C5H3F3N2O2
Molecular Weight: 180.09
Purity/Analysis Method: >95.0% (HPLC)
Form: Crystal
Color: White
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Bioscience, Nucleobases and Their Analogs
Other Product Names / Synonyms: Trifluorothymine
Formula:
C₅H₃F₃N₂O₂ MW: 180.09 g/mol Boiling Pt: 350 Melting Pt: 251…252 °C |
MDL Number:
MFCD00006024 CAS Number: 54-20-6 EINECS: 200-197-5 |
|
Specification Test Results
Purity/Analysis Method | >95.0% (HPLC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...