Tetraoctylammonium bromide ≥98.0%
Supplier: TCI America
Synonyms:
Tetra-n-octylammonium bromide
T1603-10G
T1603-25G
TCT1603-010GEA
22
USD
TCT1603-010G
TCT1603-025G
Tetraoctylammonium bromide ≥98.0%
Tetraoctylammonium bromide
CAS Number: 14866-33-2
MDL Number: MFCD00011863
Molecular Formula: C32H68BrN
Molecular Weight: 546.81
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 97
Formula:
C₃₂H₆₈BrN MW: 546.8 g/mol Melting Pt: 96 Flash Pt: 100 °C |
MDL Number:
MFCD00011863 CAS Number: 14866-33-2 EINECS: 238-936-9 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...