1,2,4-Trifluoro-5-nitrobenzene ≥97.0%
Supplier: TCI America
Synonyms:
2,4,5-Trifluoronitrobenzene
T1589-5G
T1589-25G
TCT1589-005GEA
17
USD
TCT1589-005G
TCT1589-025G
1,2,4-Trifluoro-5-nitrobenzene ≥97.0%
1,2,4-Trifluoro-5-nitrobenzene
CAS Number: 2105-61-5
MDL Number: MFCD00007051
Molecular Formula: C6H2F3NO2
Molecular Weight: 177.08
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Pale Yellow
Boiling point (°C): 195
Flash Point (°C): 89
Specific Gravity (20/20): 1.56
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1,2,4-Trifluoro-5-nitrobenzene
Formula:
C₆H₂F₃NO₂ MW: 177.08 g/mol Boiling Pt: 195 °C Melting Pt: –11 °C Density: 1.56 (20 °C) Flash Pt: 89 °C |
MDL Number:
MFCD00007051 CAS Number: 2105-61-5 EINECS: 218-281-5 UN: 2810 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...