Ethyl (trimethylsilyl)acetate ≥93.0%

Supplier: TCI America

Synonyms: Ethyl 2-(trimethylsilyl)acetate, (Trimethylsilyl)acetic acid ethyl ester

T1584-5G T1584-25G
TCT1584-005GEA 56 USD
TCT1584-005G TCT1584-25G
Ethyl (trimethylsilyl)acetate ≥93.0%
Ethyl (trimethylsilyl)acetate

CAS Number: 4071-88-9
MDL Number: MFCD00009172
Molecular Formula: C7H16O2Si
Molecular Weight: 160.29
Purity/Analysis Method: >93.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 159
Flash Point (°C): 35
Specific Gravity (20/20): 0.88


Product Categories: Synthetic Organic Chemistry, Silicon Compounds (for Synthesis)
Other Product Names / Synonyms: (Trimethylsilyl)acetic Acid Ethyl Ester, ETSA

Formula: C₇H₁₆O₂Si
MW: 160.29 g/mol
Boiling Pt: 159 °C
Density: 0.88 (20 °C)
Flash Pt: 35 °C
MDL Number: MFCD00009172
CAS Number: 4071-88-9
EINECS: 223-783-2
UN: 3272
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1de3fd99a8bbe27d08181529e2164c0347b7f196&vid=1de3fd99a8bbe27d08181529e2164c0347b7f196&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >93.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR