3-Thiopheneethanol ≥98.0%

Supplier: TCI America

Synonyms: Thiophen-3-ethanol, 2-(Thiophen-3-yl)ethanol, 2-(3-Thienyl)ethanol

T1475-25G T1475-5G
TCT1475-25GEA 333 USD
TCT1475-25G TCT1475-005G
3-Thiopheneethanol ≥98.0%
3-Thiopheneethanol

CAS Number: 13781-67-4
MDL Number: MFCD00009766
Molecular Formula: C6H8OS
Molecular Weight: 128.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 111
Flash Point (°C): 92
Specific Gravity (20/20): 1.16


Product Categories: Materials Chemistry, Conducting Polymer Research Reagents
Other Product Names / Synonyms: 3-(2-Hydroxyethyl)thiophene, 2-(3-Thienyl)ethanol

Formula: C₆H₈OS
MW: 128.19 g/mol
Boiling Pt: 111 °C
Density: 1.16 (20 °C)
Flash Pt: 92 °C
MDL Number: MFCD00009766
CAS Number: 13781-67-4
EINECS: 237-434-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c4a4c0945d08e1c33000840b9b0325c0dcfb39c&vid=6c4a4c0945d08e1c33000840b9b0325c0dcfb39c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR