2,3,6-Trifluorobenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

T1456-5G T1456-25G
TCT1456-005GEA 120 USD
TCT1456-005G TCT1456-025G
2,3,6-Trifluorobenzoic acid ≥98.0% (by GC, titration analysis)
2,3,6-Trifluorobenzoic acid

CAS Number: 2358-29-4
MDL Number: MFCD00002409
Molecular Formula: C7H3F3O2
Molecular Weight: 176.09
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 129

Formula: C₇H₃F₃O₂
MW: 176.09 g/mol
Boiling Pt: 237 °C (1013 hPa)
Melting Pt: 129 °C
MDL Number: MFCD00002409
CAS Number: 2358-29-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=afb073a170d4da1f7124653c92fd50282a922c95&vid=afb073a170d4da1f7124653c92fd50282a922c95&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR