ω,2,4-Trichloroacetophenone ≥98.0%

Supplier: TCI America

Synonyms: 2,4-Dichlorophenacyl chloride, α,2,4-Trichloroacetophenone, 2,2',4'-Trichloroacetophenone

T1405-25G T1405-250G
TCT1405-025GEA 41.8 USD
TCT1405-025G TCT1405-250G
ω,2,4-Trichloroacetophenone ≥98.0%
ω,2,4-Trichloroacetophenone

CAS Number: 4252-78-2
MDL Number: MFCD00000934
Molecular Formula: C8H5Cl3O
Molecular Weight: 223.48
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Flash Point (°C): 110


Other Product Names / Synonyms: 2,4-Dichlorophenacyl Chloride

Formula: C₈H₅Cl₃O
MW: 223.49 g/mol
Boiling Pt: 292 °C (1013 hPa)
Melting Pt: 58 - 60 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000934
CAS Number: 4252-78-2
EINECS: 224-218-2
UN: 2923
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ffb01edb9b6f56b523b43dc1e076e8261f4cbe58&vid=ffb01edb9b6f56b523b43dc1e076e8261f4cbe58&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR