2,3,4-Trifluoroaniline ≥98.0%

Supplier: TCI America
Danger

T1385-25G T1385-5G
TCT1385-025GEA 116 USD
TCT1385-025G TCT1385-005G
2,3,4-Trifluoroaniline ≥98.0%
2,3,4-Trifluoroaniline

CAS Number: 3862-73-5
MDL Number: MFCD00011737
Molecular Formula: C6H4F3N
Molecular Weight: 147.10
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 92
Flash Point (°C): 68
Specific Gravity (20/20): 1.41

Formula: C₆H₄F₃N
MW: 147.1 g/mol
Boiling Pt: 92 °C
Melting Pt: 14…15 °C
Density: 1.41 (20 °C)
Flash Pt: 68 °C
MDL Number: MFCD00011737
CAS Number: 3862-73-5
EINECS: 407-170-9
UN: 3082
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bb0b840f556f602f97bfe76a045305252f2941a4&vid=d90d402378191ad7b616364cf73b686abf6b5270&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR